Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227853.2 | XP_006227915.1 | 11, 12 | 1500 | ||
XM_008758714.1 | XP_008756936.1 | 10, 11 | 1486 | |||
XM_008758715.1 | XP_008756937.1 | 12, 13 | 1553 | |||
XM_008758716.1 | XP_008756938.1 | 12, 13 | 1551 |