Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144955.1 | NP_001138427.1 | 3, 4 | 556 | ||
NM_020234.5 | NP_064619.2 | 4, 5 | 684 | |||
XM_011521815.1 | XP_011520117.1 | 3, 4 | 605 | |||
XM_017022419.1 | XP_016877908.1 | 3, 4 | 603 | |||
XM_017022420.1 | XP_016877909.1 | 4, 5 | 1465 | |||
XM_017022421.1 | XP_016877910.1 | 4, 5 | 696 | |||
XM_017022422.1 | XP_016877911.1 | 3, 4 | 1337 | |||
XM_017022423.1 | XP_016877912.1 | 3, 4 | 605 | |||
XM_017022424.1 | XP_016877913.1 | 4, 5 | 676 | |||
XM_017022425.1 | XP_016877914.1 | 4, 5 | 676 | |||
XM_017022426.1 | XP_016877915.1 | 3, 4 | 1337 | |||
XM_017022427.1 | XP_016877916.1 | 4, 5 | 621 | |||
XM_017022428.1 | XP_016877917.1 | 4, 5 | 626 | |||
XM_017022429.1 | XP_016877918.1 | 4, 5 | 1572 | |||
XM_017022430.1 | XP_016877919.1 | 4, 5 | 775 | |||
XM_017022431.1 | XP_016877920.1 | 3, 4 | 217 | |||
GenBank mRNA | AF168717.1 | - | 3, 4 | 534 | ||
AK290184.1 | - | 3, 4 | 525 | |||
BC032535.2 | - | 4, 5 | 624 | 5527713 | ||
BC093073.1 | - | 4, 5 | 614 | 30326731 |