Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020242.2 | NP_064627.1 | 3 | 227 | ||
XM_011533964.2 | XP_011532266.1 | 3 | 230 | |||
XM_017006884.1 | XP_016862373.1 | 3 | 230 | |||
XM_017006885.1 | XP_016862374.1 | 3 | 157 | |||
XM_017006886.1 | XP_016862375.1 | 4 | 278 | |||
XM_017006887.1 | XP_016862376.1 | 3 | 230 | |||
XM_017006889.1 | XP_016862378.1 | 3 | 230 | |||
GenBank mRNA | AB035898.1 | - | 3 | 151 | ||
AK307674.1 | - | 3 | 146 | |||
BC117174.1 | - | 2 | 80 | 40125725 | ||
BC143752.1 | - | 2 | 80 | |||
DR005852.1 | - | 3 | 233 |