Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034006.1 | NP_001029178.1 | 7 | 827 | ||
XM_006248487.2 | XP_006248549.1 | 7 | 841 | |||
XM_006248489.2 | XP_006248551.1 | 7 | 840 | |||
XM_006248491.2 | XP_006248553.1 | 7 | 839 | |||
GenBank mRNA | BC090073.1 | - | 7 | 827 | 7114273 |