Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aarsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034109.1 | NP_001029281.1 | 10 | 978 | ||
GenBank mRNA | BC083790.1 | - | 10 | 978 | 7106366 | |
FQ213285.1 | - | 10 | 979 |