Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020244.2 | NP_064629.2 | 1, 2 | 504 | ||
XM_011538574.1 | XP_011536876.1 | 1, 2 | 471 | |||
GenBank mRNA | AF047431.1 | - | 1, 2 | 75 | ||
AF195623.1 | - | 1, 2 | 451 | |||
AF195624.1 | - | 1, 2 | 451 | |||
AK075211.1 | - | 1, 2 | 471 | |||
AK093067.1 | - | 2, 3 | 363 | |||
AK226127.1 | - | 1, 2 | 178 | |||
BC020819.1 | - | 1, 2 | 458 | 4696613 | ||
BC050429.1 | - | 1, 2 | 497 | 6473945 | ||
CD300296.1 | - | 1, 2 | 530 |