Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034912.1 | NP_001030084.1 | 878 | |||
GenBank mRNA | AY310142.1 | - | 267 | |||
AY321329.1 | - | 262 | ||||
BC089887.1 | - | 878 | 7371502 |
Gene Symbol: LOC103689968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232083.2 | XP_006232145.1 | 6 | 555 | ||
XM_006232084.2 | XP_006232146.1 | 5 | 921 | |||
XM_008760821.1 | XP_008759043.1 | 7 | 972 |