Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031025.1 | NP_112287.1 | 6 | 796 | ||
GenBank mRNA | BC107440.1 | - | 6 | 796 | 7115402 | |
D00092.1 | - | 3 | 300 | |||
D10655.1 | - | 5 | 513 | |||
M16075.1 | - | 3 | 300 |