Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053363.2 | NP_445815.2 | 9 | 1465 | ||
XM_006253070.2 | XP_006253132.1 | 9 | 1621 | |||
XM_006253071.2 | XP_006253133.1 | 8 | 1569 | |||
XM_006253073.2 | XP_006253135.1 | 8 | 4545 | |||
XM_006253075.2 | XP_006253137.1 | 8 | 1569 | |||
XM_006253076.2 | XP_006253138.1 | 8 | 2461 | |||
XM_006253077.2 | XP_006253139.1 | 8 | 1567 | |||
GenBank mRNA | AF142778.1 | - | 9 | 1465 | ||
D17521.1 | - | 8 | 1584 |