Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037353.1 | NP_001032430.1 | 3, 4 | 350 | ||
GenBank mRNA | BC105773.1 | - | 3, 4 | 350 | 7930490 |