Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc41a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037492.1 | NP_001032569.1 | 4 | 708 | ||
XM_006236871.2 | XP_006236933.1 | 5 | 718 | |||
XM_006236872.2 | XP_006236934.1 | 5 | 775 | |||
XM_006236873.1 | XP_006236935.1 | 5 | 1092 | |||
XM_006236874.2 | XP_006236936.1 | 4 | 647 | |||
XM_006236875.2 | XP_006236937.1 | 5 | 924 | |||
XM_006236876.2 | XP_006236938.1 | 4 | 319 | |||
XM_006236877.2 | XP_006236939.1 | 4 | 1064 | |||
XM_006236878.2 | XP_006236940.1 | 4 | 365 | |||
XM_006236879.2 | XP_006236941.1 | 4 | 360 | |||
XM_008763134.1 | XP_008761356.1 | 5 | 919 | |||
GenBank mRNA | BC104698.1 | - | 4 | 708 | 7134770 |