Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128610.2 | NP_001122082.1 | 3 | 519 | ||
NM_005154.4 | NP_005145.3 | 3 | 379 | |||
XM_006720761.3 | XP_006720824.1 | 4 | 699 | |||
XM_006720762.3 | XP_006720825.1 | 4 | 699 | |||
XM_011522193.2 | XP_011520495.1 | 4 | 536 | |||
XM_017022718.1 | XP_016878207.1 | 3 | 374 | |||
XM_017022719.1 | XP_016878208.1 | 3 | 491 | |||
XM_017022720.1 | XP_016878209.1 | 4 | 536 | |||
XM_017022721.1 | XP_016878210.1 | 3 | 362 | |||
GenBank mRNA | AK292400.1 | - | 3 | 374 | ||
BC015545.1 | - | 4 | 653 | |||
BC026954.1 | - | 3 | 469 | |||
BC038801.1 | - | 3 | 367 | |||
BC051345.1 | - | 3 | 418 | |||
BC062694.1 | - | 2 | 210 | |||
BC092443.1 | - | 3 | 341 | |||
BC110590.2 | - | 3 | 345 | 40027270 | ||
BX537420.1 | - | 3 | 374 | |||
D29956.1 | - | 3 | 493 | |||
DA112964.1 | - | 3 | 380 |