Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037509.1 | NP_001032598.1 | 2 | 113 | ||
XM_006253374.2 | XP_006253436.1 | 2 | 221 | |||
GenBank mRNA | AY621341.1 | - | 2 | 113 |