Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037523.1 | NP_001032612.1 | 1, 2 | 117 | ||
XM_006253375.2 | XP_006253437.1 | 2, 3 | 241 | |||
GenBank mRNA | AY621362.1 | - | 1, 2 | 117 |