Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040194.1 | NP_001035284.1 | 5 | 708 | ||
NM_001040195.1 | NP_001035285.1 | 4 | 673 | |||
NM_001040196.1 | NP_001035286.1 | 6 | 805 | |||
NM_001040197.1 | NP_001035287.1 | 6 | 826 | |||
NM_020350.4 | NP_065083.3 | 5 | 729 | |||
XM_011541799.2 | XP_011540101.2 | 6 | 722 | |||
XM_011541800.2 | XP_011540102.1 | 6 | 743 | |||
XM_011541802.1 | XP_011540104.1 | 7 | 819 | |||
GenBank mRNA | AF312374.1 | - | 5 | 660 | ||
AK290042.1 | - | 5 | 672 | |||
BC011946.1 | - | 5 | 614 | |||
BC017328.2 | - | 5 | 646 | 4546585 | ||
BQ672962.1 | - | 1 | 213 |