Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bglap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA016947.1 | - | 4 | 421 | ||
AK140913.1 | - | 6 | 1875 | |||
BB148904.1 | - | 1 | 103 | |||
BC019774.1 | - | 7 | 856 | |||
BC031815.1 | - | 4 | 461 | 4166503 | ||
BC055868.1 | - | 4 | 421 | 4190595 | ||
BC069910.1 | - | 639 | ||||
BC100687.1 | - | 263 | 40041102 | |||
BC101948.1 | - | 263 | 40041107 | |||
BC141238.1 | - | 288 | 9055850 | |||
BC145564.1 | - | 288 | ||||
BC145565.1 | - | 288 | 9054130 | |||
BE625312.1 | - | 4 | 426 | |||
BY414395.1 | - | 215 | ||||
BY430398.1 | - | 1 | 220 |