Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029586.2 | NP_083862.1 | 4 | 505 | ||
NM_029606.3 | NP_083882.3 | 27 | 3052 | |||
NM_145688.2 | NP_663721.2 | 6 | 727 | |||
GenBank mRNA | AK005611.1 | - | 4 | 506 | ||
AK006504.1 | - | 6 | 713 | |||
BC005723.1 | - | 6 | 539 | |||
BC048677.1 | - | 6 | 699 | |||
BC086808.1 | - | 21 | 2292 | 30632964 |