Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128304.1 | NP_001121776.1 | 6 | 538 | ||
NM_001128305.1 | NP_001121777.1 | 4 | 500 | |||
NM_001128306.1 | NP_001121778.1 | 4 | 412 | |||
NM_001177304.1 | NP_001170775.1 | 3 | 384 | |||
NM_020353.2 | NP_065086.2 | 4 | 412 | |||
XM_005247654.2 | XP_005247711.1 | 5 | 439 | |||
XM_005247655.2 | XP_005247712.1 | 5 | 356 | |||
XM_011513031.2 | XP_011511333.1 | 4 | 298 | |||
XM_017006921.1 | XP_016862410.1 | 5 | 439 | |||
GenBank mRNA | AF199023.1 | - | 4 | 316 | ||
AK128442.1 | - | 3 | 284 | |||
AK290214.1 | - | 4 | 440 | |||
AK292815.1 | - | 6 | 438 | |||
AK312809.1 | - | 4 | 354 | |||
AL833760.1 | - | 4 | 326 | |||
AY550971.1 | - | 4 | 266 | |||
BC028354.2 | - | 4 | 412 | 4831340 | ||
BI598345.1 | - | 4 | 508 | |||
BP201782.1 | - | 4 | 454 | |||
DA185957.1 | - | 6 | 502 | |||
DA444235.1 | - | 4 | 338 |