Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271386.1 | NP_001258315.1 | 5 | 337 | ||
NM_024124.3 | NP_077038.2 | 3 | 396 | |||
GenBank mRNA | AF235053.1 | - | 2 | 65 | ||
AF279371.1 | - | 2 | 65 | |||
AK087563.1 | - | 5 | 686 | |||
AK133334.1 | - | 5 | 337 | |||
BC098187.1 | - | 3 | 381 | 5363624 | ||
EL604734.1 | - | 3 | 396 |