Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054053.4 | NP_473394.3 | 20 | 4350 | ||
GenBank mRNA | AF405692.1 | - | 16 | 4495 | ||
AF405693.1 | - | 10 | 2149 | |||
AF435926.1 | - | 20 | 4351 |