Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029993.1 | NP_001025164.1 | 19 | 3767 | ||
NM_001029994.1 | NP_001025165.1 | 18 | 3695 | |||
GenBank mRNA | AK004428.1 | - | 1 | 723 | ||
AK131189.1 | - | 16 | 2838 | |||
AK159881.1 | - | 18 | 3638 | |||
AK169952.1 | - | 8 | 4756 | |||
BC030495.1 | - | 18 | 3675 | 5400494 |