Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164114.1 | NP_001157586.1 | 2 | 356 | ||
NM_001164115.1 | NP_001157587.1 | 2 | 356 | |||
NM_001164116.1 | NP_001157588.1 | 3 | 380 | |||
NM_020356.3 | NP_065089.2 | 2 | 356 | |||
XM_006723831.3 | XP_006723894.1 | 3 | 942 | |||
GenBank mRNA | AJ276678.1 | - | 3 | 380 | ||
AK027760.1 | - | 2 | 353 | |||
AK295613.1 | - | 2 | 221 | |||
AK308057.1 | - | 2 | 357 | |||
AK314653.1 | - | 2 | 289 | |||
BC027951.1 | - | 2 | 306 | 5205865 | ||
BX280195.1 | - | 2 | 306 | |||
DA584687.1 | - | 2 | 357 |