Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290541.1 | NP_001277470.1 | 5 | 1203 | ||
NM_139306.3 | NP_647467.1 | 6 | 1341 | |||
GenBank mRNA | AF282864.1 | - | 6 | 1300 | ||
AK035314.1 | - | 1 | 237 | |||
BC059819.1 | - | 5 | 1203 | 6839525 |