Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 15, 16 | 2138 | ||
GenBank mRNA | AK088950.1 | - | 14, 15 | 3728 | ||
AK129220.1 | - | 14, 15 | 3729 | |||
AK134073.1 | - | 3, 4 | 1384 | |||
BC026492.1 | - | 8, 9 | 1083 | 4505095 | ||
BC092240.1 | - | 14, 15 | 2049 | 30662573 |