Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320397.1 | NP_001307326.1 | 4 | 397 | ||
NM_001320398.1 | NP_001307327.1 | 4 | 493 | |||
NM_001320399.1 | NP_001307328.1 | 5 | 622 | |||
NM_001320401.1 | NP_001307330.1 | 3 | 421 | |||
NM_020357.2 | NP_065090.1 | 4 | 496 | |||
NR_135223.1 | 4 | 444 | ||||
NR_135224.1 | 3 | 281 | ||||
NR_135225.1 | 4 | 441 | ||||
NR_135227.1 | 4 | 697 | ||||
NR_135229.1 | 4 | 625 | ||||
NR_135230.1 | 3 | 369 | ||||
NR_135231.1 | 2 | 206 | ||||
NR_135232.1 | 5 | 539 | ||||
GenBank mRNA | AB037675.1 | - | 4 | 392 | ||
AK025551.1 | - | 4 | 336 | |||
AK098760.1 | - | 4 | 376 | |||
AK222978.1 | - | 5 | 387 | |||
AK314629.1 | - | 4 | 377 | |||
BC107796.1 | - | 4 | 338 | |||
BF541732.1 | - | 4 | 389 | |||
BI547939.1 | - | 4 | 609 | |||
BI913146.1 | - | 4 | 520 | |||
CN482876.1 | - | 5 | 423 | |||
CR627371.1 | - | 3 | 182 | |||
DA304502.1 | - | 4 | 444 | |||
DA710370.1 | - | 3 | 311 | |||
DA817951.1 | - | 4 | 361 | |||
DB032930.1 | - | 3 | 220 | |||
DB060329.1 | - | 4 | 308 | |||
DB481581.1 | - | 3 | 257 | |||
HY051831.1 | - | 2 | 94 |