Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp17lc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010089.3 | NP_034219.3 | 1473 | |||
GenBank mRNA | U70369.1 | - | 1473 |