Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020362.4 | NP_065095.2 | 3 | 406 | ||
XM_011541804.1 | XP_011540106.1 | 2 | 331 | |||
GenBank mRNA | AF218024.1 | - | 3 | 387 | ||
AF221595.1 | - | 3 | 398 | |||
AF271784.1 | - | 3 | 402 | |||
AK296662.1 | - | 2 | 331 | |||
AK310330.1 | - | 3 | 353 | |||
AK313005.1 | - | 3 | 374 | |||
BC017208.2 | - | 3 | 367 | 3943360 | ||
DB104988.1 | - | 3 | 406 |