Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134082.3 | NP_598843.3 | 16 | 2124 | ||
GenBank mRNA | BC030329.1 | - | 9 | 1112 | ||
BC141229.1 | - | 15 | 1685 | |||
DV655111.1 | - | 4 | 485 |