Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025888.5 | NP_080164.2 | 8 | 1502 | ||
GenBank mRNA | AK010404.1 | - | 7 | 1265 | ||
AK013254.1 | - | 3 | 632 | |||
AK030202.1 | - | 8 | 1504 | |||
AK037247.1 | - | 1 | 101 | |||
AK088807.1 | - | 7 | 1270 | |||
AK166987.1 | - | 3 | 641 | |||
AK168440.1 | - | 8 | 1493 | |||
AK171742.1 | - | 7 | 1300 | |||
BC036318.1 | - | 6 | 1147 | |||
BC048161.1 | - | 7 | 1286 | |||
BC049250.1 | - | 7 | 1379 | 5042309 | ||
BC052712.1 | - | 7 | 1288 |