Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133766.3 | NP_598527.2 | 13 | 1543 | ||
GenBank mRNA | AB158474.1 | - | 13 | 1547 | ||
AK018695.1 | - | 8 | 762 | |||
AK030074.1 | - | 13 | 1544 | |||
AK032461.1 | - | 13 | 1516 | |||
AK044000.1 | - | 13 | 1537 | |||
AK044475.1 | - | 13 | 1539 | |||
AK129065.1 | - | 13 | 1521 | |||
AK161250.1 | - | 13 | 1528 | |||
BC007482.1 | - | 13 | 1547 | 3495608 | ||
BC032092.1 | - | 13 | 1548 |