Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024241.2 | NP_077203.2 | 7 | 1474 | ||
GenBank mRNA | AK050683.1 | - | 7 | 1460 | ||
AK084967.1 | - | 4 | 1073 | |||
AK135307.1 | - | 7 | 1475 | |||
BC067395.1 | - | 7 | 1466 | 30460089 |