Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024241.2 | NP_077203.2 | 11 | 3564 | ||
GenBank mRNA | AK084967.1 | - | 8 | 3163 | ||
BC067395.1 | - | 11 | 3556 | 30460089 |