Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010495.3 | NP_034625.1 | 1 | 335 | ||
GenBank mRNA | AB221668.1 | - | 1 | 245 | ||
AK008264.1 | - | 1 | 332 | |||
BC025073.1 | - | 1 | 312 | 4206508 | ||
CT010243.1 | - | 1 | 245 | |||
M31885.1 | - | 1 | 331 | |||
U43884.1 | - | 1 | 253 |