Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020375.2 | NP_065108.1 | 2, 3 | 157 | ||
GenBank mRNA | AJ272206.1 | - | 2, 3 | 145 | ||
AK313226.1 | - | 2, 3 | 130 | |||
AY425618.1 | - | 2, 3 | 116 | |||
BC012340.1 | - | 2, 3 | 116 | 4576725 | ||
DB480618.1 | - | 2, 3 | 160 |