Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5830411N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128145.1 | NP_001121617.1 | 8 | 2007 | ||
NM_001128146.1 | NP_001121618.1 | 11 | 2358 | |||
NM_175533.3 | NP_780742.1 | 5 | 2384 | |||
GenBank mRNA | AK030813.1 | - | 7 | 3191 | ||
AK040624.1 | - | 5 | 2385 | |||
AK220311.1 | - | 5 | 3145 | |||
EF624462.1 | - | 11 | 2358 | |||
EF624463.1 | - | 8 | 2010 |