Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040106.2 | NP_001035195.1 | 21 | 4824 | ||
NM_177762.6 | NP_808430.2 | 20 | 4581 | |||
GenBank mRNA | AK129272.1 | - | 11 | 3042 | ||
AK147363.1 | - | 21 | 4825 | |||
BC043125.1 | - | 10 | 2630 | |||
BC141176.1 | - | 21 | 4580 | 9055788 |