Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1b27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009596.1 | NP_033726.1 | 2, 3 | 243 | ||
GenBank mRNA | AF039064.1 | - | 1, 2 | 176 | ||
BC016132.1 | - | 2, 3 | 243 | 4922791 | ||
BC132436.1 | - | 2, 3 | 327 | 40130713 | ||
GU269231.1 | - | 2, 3 | 242 |