Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011654.2 | NP_035784.1 | 4 | 1410 | ||
GenBank mRNA | AK075955.1 | - | 4 | 1457 | ||
AK137885.1 | - | 4 | 1454 | |||
AK150128.1 | - | 4 | 1457 | |||
AK150179.1 | - | 4 | 1457 | |||
AK150968.1 | - | 4 | 1456 | |||
AK151199.1 | - | 4 | 1454 | |||
AK151809.1 | - | 4 | 1454 | |||
AK152298.1 | - | 4 | 1454 | |||
AK153064.1 | - | 4 | 1454 | |||
AK153254.1 | - | 4 | 1454 | |||
AK158958.1 | - | 4 | 1586 | |||
AK164428.1 | - | 4 | 1457 | |||
AK168770.1 | - | 4 | 1454 | |||
AK169075.1 | - | 4 | 1454 | |||
AK169092.1 | - | 4 | 1457 | |||
AK169130.1 | - | 4 | 1457 | |||
AK169665.1 | - | 4 | 1457 | |||
AK191172.1 | - | 1 | 264 | |||
AK192129.1 | - | 1 | 243 | |||
AK192775.1 | - | 1 | 261 | |||
AK193004.1 | - | 1 | 222 | |||
AK194180.1 | - | 1 | 222 | |||
AK194222.1 | - | 1 | 216 | |||
AK196308.1 | - | 1 | 231 | |||
AK197610.1 | - | 1 | 264 | |||
AK198269.1 | - | 1 | 264 | |||
AK199828.1 | - | 1 | 211 | |||
AK200029.1 | - | 1 | 235 | |||
AK200084.1 | - | 1 | 264 | |||
AK203041.1 | - | 1 | 187 | |||
AK207058.1 | - | 1 | 221 | |||
AK211470.1 | - | 1 | 232 | |||
BC002219.1 | - | 4 | 1444 | 3490568 | ||
BC004790.1 | - | 2 | 1045 | |||
BC008117.1 | - | 4 | 1437 | 3498312 | ||
BC020390.1 | - | 1297 | ||||
BC063777.1 | - | 4 | 1407 | 4458530 | ||
BC083120.1 | - | 4 | 1435 | 6311394 | ||
BC098321.1 | - | 4 | 1437 | 5065638 | ||
BC108337.1 | - | 4 | 1436 | 4167097 | ||
BC108394.1 | - | 4 | 1410 | 6505244 | ||
M13446.1 | - | 4 | 1434 | |||
M28727.1 | - | 2 | 1058 |