Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146663.2 | NP_666874.2 | 1 | 830 | ||
GenBank mRNA | BC116806.1 | - | 1 | 830 | 40126125 | |
BC116808.1 | - | 1 | 830 | 40126127 |