Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC117002.1 | - | 1 | 379 | 40126321 | |
BC120742.1 | - | 1 | 379 | 40129665 |