Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146441.1 | NP_666652.1 | 1 | 596 | ||
GenBank mRNA | BC119155.1 | - | 1 | 623 | 8733904 | |
BC119157.1 | - | 1 | 623 | 8733906 |