Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301769.1 | NP_001288698.1 | 3, 4 | 603 | ||
NM_001301771.1 | NP_001288700.1 | 2, 3 | 572 | |||
NM_001301773.1 | NP_001288702.1 | 2, 3 | 679 | |||
NM_001301774.1 | NP_001288703.1 | 2, 3 | 572 | |||
NM_001301775.1 | NP_001288704.1 | 3, 4 | 644 | |||
NM_020422.5 | NP_065155.3 | 2, 3 | 679 | |||
NR_125971.1 | 3, 4 | 644 | ||||
XM_005255439.4 | XP_005255496.1 | 3, 4 | 331 | |||
XM_006721066.2 | XP_006721129.1 | 3, 4 | 779 | |||
XM_006721067.3 | XP_006721130.1 | 3, 4 | 779 | |||
XM_006721069.3 | XP_006721132.1 | 3, 4 | 779 | |||
XM_017023471.1 | XP_016878960.1 | 3, 4 | 779 | |||
XM_017023472.1 | XP_016878961.1 | 3, 4 | 779 | |||
XM_017023473.1 | XP_016878962.1 | 2, 3 | 415 | |||
XM_017023474.1 | XP_016878963.1 | 2, 3 | 415 | |||
GenBank mRNA | AF070596.1 | - | 3, 4 | 644 | ||
AK074667.1 | - | 2, 3 | 377 | |||
AK293557.1 | - | 3, 4 | 332 | |||
AY212918.1 | - | 1, 2 | 104 | |||
BC015812.1 | - | 1, 2 | 216 | 3920903 | ||
BI821683.1 | - | 3, 4 | 275 | |||
CB128337.1 | - | 2, 3 | 288 | |||
CX786237.1 | - | 2, 3 | 363 | |||
DA893031.1 | - | 2, 3 | 394 | |||
DB054887.1 | - | 2, 3 | 358 |