Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020196.2 | NP_064581.2 | 2 | 189 | ||
GenBank mRNA | AB026111.1 | - | 2 | 178 | ||
AF226051.1 | - | 2 | 189 | |||
AF258567.1 | - | 2 | 184 | |||
AF272147.1 | - | 2 | 199 | |||
AK074035.1 | - | 1 | 84 | |||
BC007208.1 | - | 2 | 171 | 2823235 |