Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020197.2 | NP_064582.2 | 2 | 222 | ||
GenBank mRNA | AF226053.1 | - | 2 | 211 | ||
AK313868.1 | - | 2 | 192 | |||
BC098133.1 | - | 2 | 211 | 40005281 | ||
BC098276.1 | - | 2 | 211 | 40005283 | ||
BC098305.1 | - | 2 | 211 | 40005284 | ||
BC098335.1 | - | 2 | 211 | 40005280 | ||
BG723585.1 | - | 2 | 349 |