Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201363.2 | NP_958751.2 | 2 | 97 | ||
GenBank mRNA | AK003650.1 | - | 2 | 98 | ||
AY367775.1 | - | 1 | 27 | |||
BC132091.1 | - | 1 | 49 | 40130368 |