Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1313 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207150.1 | NP_997033.1 | 1 | 521 | ||
GenBank mRNA | BC127964.1 | - | 1 | 525 | 40134976 |