Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmsb15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207267.4 | NP_997150.1 | 4 | 508 | ||
GenBank mRNA | AK029707.1 | - | 4 | 485 | ||
BB623661.1 | - | 4 | 504 | |||
BC096054.1 | - | 4 | 441 | 3373135 | ||
BE625563.1 | - | 4 | 433 |
Gene Symbol: Tmsb15b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081983.1 | NP_001075452.1 | 3 | 403 |
Gene Symbol: Tmsb15b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080967.4 | NP_001074436.1 | 3 | 368 | ||
GenBank mRNA | CN836907.1 | - | 354 | |||
CN842750.1 | - | 3 | 371 |