Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175656.3 | NP_783587.1 | 1 | 360 | ||
GenBank mRNA | AK011560.1 | - | 1 | 361 | ||
BC019757.1 | - | 1 | 350 | |||
BC052219.1 | - | 1 | 333 | |||
BC092144.1 | - | 1 | 350 | 30612926 | ||
BC125598.1 | - | 1 | 344 | 40130113 | ||
BC125600.1 | - | 1 | 344 | 40130115 |