Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005269.2 | NP_005260.1 | 3, 4 | 278 | ||
XM_011538189.2 | XP_011536491.1 | 3, 4 | 524 | |||
XM_011538190.1 | XP_011536492.1 | 3, 4 | 291 | |||
GenBank mRNA | AB239135.1 | - | 2, 3 | 117 | ||
BC013000.2 | - | 2, 3 | 238 | 3531657 | ||
X07384.1 | - | 3, 4 | 278 |