Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133203.5 | NP_573466.3 | 4 | 365 | ||
GenBank mRNA | AB033769.1 | - | 4 | 378 | ||
AB190779.1 | - | 3 | 224 | |||
AB190780.1 | - | 3 | 224 | |||
AB190781.1 | - | 3 | 224 | |||
AB190782.1 | - | 3 | 224 | |||
AB190783.1 | - | 3 | 224 | |||
AB190784.1 | - | 3 | 224 | |||
AB190785.1 | - | 3 | 224 | |||
AB190786.1 | - | 3 | 224 | |||
AB190787.1 | - | 3 | 224 | |||
AB190788.1 | - | 3 | 224 | |||
AB190789.1 | - | 3 | 224 | |||
AB193831.1 | - | 3 | 224 | |||
AB193832.1 | - | 3 | 224 | |||
AB193833.1 | - | 3 | 224 | |||
AK080021.1 | - | 4 | 366 | |||
AY620246.1 | - | 3 | 228 | |||
AY642595.1 | - | 3 | 228 | |||
BC132236.1 | - | 4 | 326 | 40130513 |